Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ethyl-3-(trifluoromethyl)crotonate cis- and trans-mixture ≥98%

Supplier: Matrix Scientific
Synonyms: 3-(Trifluoromethyl)crotonic acid ethyl ester

006387-5G
101779-036EA 361.54 USD InStock
101779-036
Ethyl-3-(trifluoromethyl)crotonate cis- and trans-mixture ≥98%
Ethyl-3-(trifluoromethyl)crotonate
Hazard: Flammable/Irritant


Formula: C₇H₉F₃O₂
MW: 182.14
Boiling Pt: 129…130 °C
MDL Number: MFCD00040846
CAS Number: 24490-03-7
UN: 3272
ADR: 3,III
Order Now
Specification Test Results
Boiling point 129-130°
ND20 1.3741

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty